Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552713 Supplier Invitrogen™ Supplier No. PA552713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILTSEFSYQH SLSILVEEFL QSKELRATVT QMEHHHLFSN ILDVLGASQR FFEDLEQRHK AQVLVEDISD ILEEHAEKYF HPYIAYCSNE VYQQRTLQKL ISSNAAFREA LREIERRPAC GGLPMLSFLI LPMQRVTRLP LLMDTLC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

Although the specific function of this protein is not known yet, it is thought to be involved in protein-protein and protein-lipid interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arhgef16
Gene Accession No. Q5VV41
Gene Alias Arhgef16; EPHEXIN4; Ephexin-4; GEF16; NBR; Neuroblastoma; Rho guanine exchange factor (GEF) 16; Rho guanine nucleotide exchange factor (GEF) 16; Rho guanine nucleotide exchange factor 16
Gene Symbols Arhgef16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGEF16. Recombinant protein control fragment (Product #RP-88841).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.