Learn More
Invitrogen™ ARHGEF17 Polyclonal Antibody

Description
A suggested positive control is Jurkat cell lysate. PA5-20440 can be used with blocking peptide PEP-0557.
Specifications
Specifications
| Antigen | ARHGEF17 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ARHGEF17 |
| Gene Accession No. | Q96PE2 |
| Gene Alias | 164 kDa Rho-specific guanine-nucleotide exchange factor; 8030463K16; AI428794; Arhgef17; AW558066; BC035332; Kiaa0337; mKIAA0337; P164RHOGEF; p164-RhoGEF; Rho guanine nucleotide exchange factor (GEF) 17; Rho guanine nucleotide exchange factor 17; Rho-specific guanine-nucleotide exchange factor 164 kDa; TEM4; Tumor endothelial marker 4 |
| Gene Symbols | ARHGEF17 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.