Learn More
Invitrogen™ ARHGEF18 Polyclonal Antibody

Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (89%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ARHGEF18 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Arhgef18 |
| Gene Accession No. | Q6P9R4, Q6ZSZ5 |
| Gene Alias | 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF |
| Gene Symbols | Arhgef18 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.