Learn More
Invitrogen™ ARHGEF3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ARHGEF3 protein.
Specifications
Specifications
| Antigen | ARHGEF3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ARHGEF3 |
| Gene Accession No. | Q91X46, Q9NR81 |
| Gene Alias | 1200004I24Rik; 59.8 kDA protein; 9830169H03Rik; ARHG3; ARHGEF3; C76747; DKFZP434F2429; Exchange factor found in platelets and leukemic and neuronal tissues; exchange factor found in platelets and leukemic and neuronal tissues, XPLN; GEF3; MGC118905; Rho guanine nucleotide exchange factor (GEF) 3; Rho guanine nucleotide exchange factor 3; RhoGEF protein; STA3; Xpln |
| Gene Symbols | ARHGEF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.