Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598942 Supplier Invitrogen™ Supplier No. PA598942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARHGEF3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARHGEF3
Gene Accession No. Q9NR81
Gene Alias 1200004I24Rik; 59.8 kDA protein; 9830169H03Rik; ARHG3; ARHGEF3; C76747; DKFZP434F2429; Exchange factor found in platelets and leukemic and neuronal tissues; exchange factor found in platelets and leukemic and neuronal tissues, XPLN; GEF3; MGC118905; Rho guanine nucleotide exchange factor (GEF) 3; Rho guanine nucleotide exchange factor 3; RhoGEF protein; STA3; Xpln
Gene Symbols ARHGEF3
Host Species Rabbit
Immunogen Recombinant Human Rho guanine nucleotide exchange factor 3 protein (287-526AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50650
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.