Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583532 Supplier Invitrogen™ Supplier No. PA583532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSPFRSINIP EPVLPSEDFT NLLPSQAYEK AQESRSVHVG DSNVKGMMGP GVNPTTPEAE ENLK Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.

May act as a guanine-nucleotide releasing factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF33
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF33
Gene Accession No. A8MVX0
Gene Alias ARHGEF33; DH and coiled-coil domain-containing protein ENSP00000381780; Rho guanine nucleotide exchange factor (GEF) 33; Rho guanine nucleotide exchange factor 33
Gene Symbols ARHGEF33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGEF33. Recombinant protein control fragment (Product #RP-105996).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100271715
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.