Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565794 Supplier Invitrogen™ Supplier No. PA565794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYQALNSLLVA ELPQFNQLVM QWLGQILCTF VTLQRDLAKQ VLQRAEGSMA QLPHHHVPEP AFRKLVEDAL GRTSNQLRSF QETFEKVQPP P Highest antigen sequence identity to the following orthologs - mouse 78%, rat 76%.

May act as a guanine nucleotide exchange factor (GEF).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF37
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF37
Gene Accession No. A1IGU5
Gene Alias 4933429F08Rik; ARHGEF37; probable guanine nucleotide exchange factor FLJ41603 homolog; protein 2-88; RGD1560471; Rho guanine nucleotide exchange factor (GEF) 37; Rho guanine nucleotide exchange factor 37; Scaffold protein tuba 3
Gene Symbols ARHGEF37
Host Species Rabbit
Immunogen Recombinant Human ARHGEF37. Recombinant protein control fragment (Product #RP-106467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.