Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565410 Supplier Invitrogen™ Supplier No. PA565410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEEEHHMKRMM AKREKIIKEL IQTEKDYLND LELCVREVVQ PLRNKKTDRL DVDSLFSNIE SVHQIS Highest antigen sequence identity to the following orthologs - mouse 78%, rat 82%.

May act as a guanine-nucleotide releasing factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF38
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF38
Gene Accession No. Q9NXL2
Gene Alias ARHGEF38; Rho guanine nucleotide exchange factor (GEF) 38; rho guanine nucleotide exchange factor 38
Gene Symbols ARHGEF38
Host Species Rabbit
Immunogen Recombinant Human ARHGEF38. Recombinant protein control fragment (Product #RP-106092).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54848
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.