Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF39 Polyclonal Antibody
ARHGEF39 Polyclonal Antibody
Supplier: Thermo Scientific PA563945
Description
ARHGEF39 Polyclonal Antibody for IHC

Specifications
ARHGEF39 | |
Polyclonal | |
Unconjugated | |
ARHGEF39 | |
ARHGEF39; C9orf100; Rho guanine nucleotide exchange factor (GEF) 39; Rho guanine nucleotide exchange factor 39; vav-like protein C9orf100 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84904 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4T4 | |
ARHGEF39 | |
Recombinant protein corresponding to Human ARHGEF39. Recombinant protein control fragment (Product #RP-101782). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction