Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF4 Antibody (ARHGEF-08), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262217DL594
Description
ARHGEF4 Monoclonal specifically detects ARHGEF4 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
ARHGEF4 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARHGEF4 | |
Recombinant fragment of human ARHGEF4 (amino acids 143-271) | |
0.1 mL | |
Primary | |
The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
ARHGEF-08 | |
Western Blot, Flow Cytometry | |
APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6 | |
Mouse | |
Protein A purified | |
Signal Transduction | |
50649 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction