Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF4 Antibody (ARHGEF-08), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262217MFV500
Description
ARHGEF4 Monoclonal antibody specifically detects ARHGEF4 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
ARHGEF4 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
ARHGEF-08 | |
Flow Cytometry, Flow (Intracellular) | |
APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6 | |
Recombinant fragment of human ARHGEF4 (amino acids 143-271) | |
0.1 mL | |
Signal Transduction | |
50649 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction