Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF4 Mouse anti-Human, Clone:ARHGEF-08, Abnova™

Catalog No. 89333052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-052 Supplier Abnova Corporation Supplier No. MAB16945
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ARHGEF4.

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene. [provided by RefSeq]

Specifications

Antigen ARHGEF4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone ARHGEF-08
Conjugate Unconjugated
Description Rho guanine nucleotide exchange factor (GEF) 4
Dilution Flow Cytometry (intracellular staining)
Western Blot
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (15 mM sodium azide)
Gene ARHGEF4
Gene Alias ASEF, ASEF1, GEF4, STM6
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 143-271 of human ARHGEF4.
Quantity 100 μg
Gene ID (Entrez) 50649
Test Specificity This antibody recognizes human intracellular protein ARHGEF4/ASEF1, approximately 80 kDa guanine nucleotide exchange factor specific for Rac1 and Cdc42.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.