Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568317 Supplier Invitrogen™ Supplier No. PA568317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arhgef4
Gene Accession No. Q7TNR9, Q9NR80
Gene Alias 9330140K16Rik; APC-stimulated guanine nucleotide exchange factor; APC-stimulated guanine nucleotide exchange factor 1; Arhgef4; Asef; ASEF1; DKFZp434G2016; ENSMUSG00000070955; GEF4; Kiaa1112; LOC100067370; LOW QUALITY PROTEIN: rho guanine nucleotide exchange factor 4; Rho guanine nucleotide exchange factor (GEF) 4; Rho guanine nucleotide exchange factor 4; STM6
Gene Symbols Arhgef4
Host Species Rabbit
Immunogen Synthesized peptide derived from human ARHGEF4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226970, 301334, 50649
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.