Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGEF6 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562890
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2890 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2890 Supplier Bethyl Laboratories, Inc Supplier No. A302558AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Acts as a RAC1 guanine nucleotide exchange factor (GEF).

Specifications

Antigen ARHGEF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ARHGEF6
Gene Accession No. Q15052
Gene Alias Rho guanine nucleotide exchange factor 6, Alpha-Pix, COOL-2, PAK-interacting exchange factor alpha, Rac/Cdc42 guanine nucleotide exchange factor 6, ARHGEF6, COOL2, KIAA0006, PIXA
Gene Symbols ARHGEF6
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9459
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.