Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117482
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117482 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117482 Supplier Invitrogen™ Supplier No. PA5117482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF7
Applications Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.1M NaCl and 15mM sodium azide; pH 7.2
Gene Arhgef7
Gene Accession No. Q14155
Gene Alias ARHGEF7; betaPix; Beta-Pix; betaPix-b; betaPix-c; Cool; COOL1; cool-1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; p85; p85Cool1; p85SPR; PAK3; Pak3 binding protein; Pak3bp; PAK-interacting exchange factor beta; PIX; Pixb; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor (GEF7); Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein
Gene Symbols Arhgef7
Host Species Rabbit
Immunogen Recombinant Human Rho guanine nucleotide exchange factor 7 protein (1-250aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8874
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.