Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHGEF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571401
Description
ARHGEF9 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a Rho-like GTPase that switches between the active state and inactive state to regulate CDC42 and other genes. Defects in this gene are a cause of startle disease with epilepsy, also known as hyperekplexia with epilepsy. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
ARHGEF9 | |
Polyclonal | |
Unconjugated | |
ARHGEF9 | |
9630036L12Rik; A230067K14; ARHDH9; ARHGEF9; Cdc42 guanine nucleotide exchange factor (GEF) 9; Cdc42 guanine nucleotide exchange factor 9; COLLYBISTIN; collybistin I; EIEE8; HPEM-2; hPEM-2 collybistin; KIAA0424; mKIAA0424; PEM2; PEM-2; PEM-2 homolog; rac/Cdc42 guanine nucleotide exchange factor 9; Rho guanine nucleotide exchange factor 9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
236915 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3UTH8 | |
ARHGEF9 | |
synthetic peptide directed towards the C terminal region of mouse ARHGEF9. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction