Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111413 Supplier Invitrogen™ Supplier No. PA5111413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPPSYPPPQD PLNHGQYLVP DGIAQSQVFE FTEPKRSQSP FWQNFSRLTP FKK Highest antigen sequence indentity to the following orthologs: Rat - 47%, Mouse - 98%.

The protein encoded by this gene is a Rho-like GTPase that switches between the active state and inactive state to regulate CDC42 and other genes. Defects in this gene are a cause of startle disease with epilepsy, also known as hyperekplexia with epilepsy. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF9
Gene Accession No. O43307
Gene Alias 9630036L12Rik; A230067K14; ARHDH9; ARHGEF9; Cdc42 guanine nucleotide exchange factor (GEF) 9; Cdc42 guanine nucleotide exchange factor 9; COLLYBISTIN; collybistin I; EIEE8; HPEM-2; hPEM-2 collybistin; KIAA0424; mKIAA0424; PEM2; PEM-2; PEM-2 homolog; rac/Cdc42 guanine nucleotide exchange factor 9; Rho guanine nucleotide exchange factor 9
Gene Symbols ARHGEF9
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ARHGEF9. Recombinant protein control fragment (Product #RP-105012).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.