Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553438 Supplier Invitrogen™ Supplier No. PA553438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQYGPQGNYS RPPAYSGVPS ASYSGPGPGM GISANNQMHG QGPSQPCGAV PLGRMPSAGM QNRPFPGNMS SMTPSSPGMS QQGGPGMGPP MPTVNRKAQE AAAAVMQAAA NSAQSR Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Feb 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID1B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARID1B
Gene Accession No. Q8NFD5
Gene Alias 6A3-5; 8030481M12; 9330189K18Rik; AI836955; Ardi1b; ARID domain-containing protein 1B; ARID1B; AT rich interactive domain 1B (Swi1 like); AT rich interactive domain 1B (SWI1-like); AT rich interactive domain 1B (SWI-like); AT-rich interaction domain 1B; AT-rich interactive domain-containing protein 1B; B230217J03Rik; BAF250B; BRG1-associated factor 250b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; BRIGHT; CSS1; DAN15; ELD (eyelid)/OSA protein; ELD/OSA1; hOsa2; KIAA1235; mKIAA1235; MRD12; Osa homolog 2; OSA2; P250R; Smcf1; smcf-1; transcription factor 1
Gene Symbols ARID1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARID1B. Recombinant protein control fragment (Product #RP-91593).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.