Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARID2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126614IR
Description
ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western BlotSpecifications
ARID2 | |
Polyclonal | |
Western Blot | |
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 | |
The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528). | |
0.1 mL | |
Chromatin Modifiers | |
196528 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction