Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551258

Catalog No. PA551258


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ARID2 protein.

ARID2 is a subunit of the PBAF chromatin-remodeling complex which facilitates ligand-dependent transcriptional activation by nuclear receptors.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARID2
Polyclonal
Unconjugated
ARID2
1700124K17Rik; 4432409D24Rik; arid 2; ARID domain-containing protein 2; ARID2; AT rich interactive domain 2 (ARID, RFX-like); AT rich interactive domain 2 (Arid-rfx like); AT-rich interaction domain 2; AT-rich interactive domain-containing protein 2; Baf200; BRG1-associated factor 200; KIAA1557; p200; RFX-like; Zinc finger protein with activation potential; zipzap; zipzap/p200
Rabbit
Antigen affinity chromatography
RUO
196528, 77044
-20°C
Liquid
Immunohistochemistry, Western Blot
0.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q68CP9
ARID2
Synthetic peptide of human ARID2.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.