Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121214 Supplier Invitrogen™ Supplier No. PA5121214
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2. The target is usually found in the following locations: Nucleus. Immunogen sequence: QNVVPQNTPM PPSPAVQVQG QPNSSQPSPF SGSSQPGDPM RKPGQNFMCL WQSCKKWFQT PSQVFYHAAT EHGGKDVYPG QCLWEGCEPF QRQRFSFITH LQDKHCSKDA LLAGLKQDEP GQAGSQKSST KQPTVGGTSS TPRAQKAIVN HPSAALMALR RGSRNLVFRD FTDEKEGPIT KHIRLTAALI LKNIGKYSEC GRRLLKRHEN NLSVLAISNM EASSTLAKCL YELNFTVQSK EQEKDSEMLQ.

ARID2 is a subunit of the PBAF chromatin-remodeling complex which facilitates ligand-dependent transcriptional activation by nuclear receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARID2
Gene Accession No. Q68CP9
Gene Alias 1700124K17Rik; 4432409D24Rik; arid 2; ARID domain-containing protein 2; ARID2; AT rich interactive domain 2 (ARID, RFX-like); AT rich interactive domain 2 (Arid-rfx like); AT-rich interaction domain 2; AT-rich interactive domain-containing protein 2; Baf200; BRG1-associated factor 200; KIAA1557; p200; RFX-like; Zinc finger protein with activation potential; zipzap; zipzap/p200
Gene Symbols ARID2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1586-1835 of human ARID2 (NP_689854.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196528
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.