Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARID3B Antibody (008), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289222B
Description
ARID3B Monoclonal specifically detects ARID3B in Human samples. It is validated for Western Blot.Specifications
ARID3B | |
Monoclonal | |
Biotin | |
ARID domain-containing protein 3B, AT rich interactive domain 3B (BRIGHT- like), AT rich interactive domain 3B (BRIGHT-like), AT-rich interactive domain-containing protein 3B, BDP, bright and dead ringer protein, DRIL2 | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human ARID3B. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
8 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
10620 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction