Learn More
Invitrogen™ ARID4A Polyclonal Antibody

Description
Recommended positive controls: HepG2. Predicted reactivity: Cow: 86%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Mouse: 92%; Pig: 86%; Rabbit: 86%; Rat: 93%.
Specifications
Specifications
| Antigen | ARID4A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ARID4A |
| Gene Accession No. | P29374 |
| Gene Alias | A630009N03; A630067N03Rik; ARID domain-containing protein 4A; ARID4A; AT rich interactive domain 4A (Rbp1 like); AT rich interactive domain 4A (RBP1-like); AT-rich interaction domain 4A; AT-rich interactive domain-containing protein 4A; MmRBBP1; Rbbp1; RBBP-1; RBP1; RBP-1; retinoblastoma binding protein 1; retinoblastoma-binding protein 1 |
| Gene Symbols | ARID4A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.