Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540646 Supplier Invitrogen™ Supplier No. PA540646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NSSKCTPVKH LNVSKPQKLA RSPARISPHI KDGEKDKHRE KHPNSSPRTY Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 86%; Rabbit: 86%; Rat: 93%.

ARID4A encodes a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein (pRB) which regulates cell proliferation. pRB represses transcription by recruiting the encoded protein. This protein, in turn, serves as a bridging molecule to recruit HDACs and, in addition, provides a second HDAC-independent repression function. The encoded protein possesses transcriptional repression activity. Multiple alternatively spliced transcripts have been observed for this gene, although not all transcript variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID4A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARID4A
Gene Accession No. P29374
Gene Alias A630009N03; A630067N03Rik; ARID domain-containing protein 4A; ARID4A; AT rich interactive domain 4A (Rbp1 like); AT rich interactive domain 4A (RBP1-like); AT-rich interaction domain 4A; AT-rich interactive domain-containing protein 4A; MmRBBP1; Rbbp1; RBBP-1; RBP1; RBP-1; retinoblastoma binding protein 1; retinoblastoma-binding protein 1
Gene Symbols ARID4A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ARID4A (aa 1119-1168).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5926
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.