Learn More
ARID4B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302233A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
The novel BRCAA1 protein′s epitope, SSKKQKRSHK, was mapped to the cytoplasm of MCF-7 cells. Immunohistochemistry analysis showed that the antigen epitope exhibited positive expression in 65% of breast cancer specimens. Its expression is associated withSpecifications
ARID4B | |
Polyclonal | |
Unconjugated | |
ARID4B | |
Breast cancer-associated antigen BRCAA1,Rb-binding protein homolog, | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51742 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q4LE39 | |
ARID4B | |
Between 125 and 175 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.