Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARID5A Monoclonal Antibody (P921)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518306
Description
Recommended positive controls: 3xFlag-human ARID5A-transfected 293T, THP-1 nuclear extract. Predicted reactivity: Mouse (83%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
Specifications
ARID5A | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q03989 | |
ARID5A | |
Recombinant fragment corresponding to a region within amino acids 1 and 259 of ARID5A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
P921 | |
Unconjugated | |
ARID5A | |
ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14 | |
Mouse | |
Protein G | |
RUO | |
10865 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction