Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARID5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543652
Description
Peptide sequence: MAAGLMHFPP TSFDSALRHR LCPASSAWHA PPVTTYAAPH FFHLNTKL Sequence homology: Cow: 80%; Dog: 100%; Guinea Pig: 86%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
Specifications
| ARID5A | |
| Polyclonal | |
| Unconjugated | |
| ARID5A | |
| ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10865 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q03989 | |
| ARID5A | |
| Synthetic peptide directed towards the C-terminal of human ARID5A (aa 547-594). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction