Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARID5A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582850
Description
Immunogen sequence: LVLPYVRHLK GEDDKPLPTS KPRKQYKMAK ENRGDDGATE RPKKAKEERR MDQMMPGKTK ADAADPAPLP SQEPPRNSTE QQGLASGSSV SFVGASGCPE AYKRLLSSFY CKGTH.
Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
Specifications
ARID5A | |
Polyclonal | |
Unconjugated | |
ARID5A | |
ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10865 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q03989 | |
ARID5A | |
Recombinant protein corresponding to Human ARID5A. Recombinant protein control fragment (Product #RP-101877). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction