Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARID5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113937 Supplier Invitrogen™ Supplier No. PA5113937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWKNVYDELG GSPGSTSAAT CTRRHYERLV LPYVRHLKGE DDKPLPTSKP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Members of the ARID protein family, including ARID5A, have diverse functions but all appear to play important roles in development, tissue-specific gene expression, and regulation of cell growth (Patsialou et al., 2005 [PubMed 15640446]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID5A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARID5A
Gene Accession No. Q03989
Gene Alias ARID domain-containing protein 5A; Arid5a; AT rich interactive domain 5A (Mrf1 like); AT rich interactive domain 5A (MRF1-like); AT-rich interaction domain 5A; AT-rich interactive domain-containing protein 5A; D430024K22Rik; hypothetical protein LOC524118; LOW QUALITY PROTEIN: AT-rich interactive domain-containing protein 5A; modulator recognition factor 1; modulator recognition factor I; MRF1; MRF-1; RFVG5814; RP11-363D14
Gene Symbols ARID5A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human ARID5A (aa 110-159).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10865
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.