Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID5B Antibody, Novus Biologicals™
SDP

Catalog No. NBP169169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169169 100 μL
NBP16916920 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP169169 Supplier Novus Biologicals Supplier No. NBP169169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARID5B Polyclonal specifically detects ARID5B in Human samples. It is validated for Western Blot.

Specifications

Antigen ARID5B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q14865
Gene Alias ARID domain-containing protein 5B, AT rich interactive domain 5B (MRF1-like), DESRTAT-rich interactive domain-containing protein 5B, FLJ21150, Modulator recognition factor 2, modulator recognition factor 2 (MRF2), MRF-2, MRF2MRF1-like protein
Gene Symbols ARID5B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ARID5B (AT rich interactive domain 5B (MRF1-like)) The peptide sequence was selected from the C terminal of ARID5B (NP_115575). Peptide sequence: VSPLDPSKEVSGKEKASEQESEGSKAAHGGHSGGGSEGHKLPLSSPIFPG.
Molecular Weight of Antigen 132 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84159
Test Specificity Guinea pig: 86%.
Reconstitution Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL.
Target Species Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.