Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107065 Supplier Thermo Scientific Supplier No. PA5107065
Add to Cart
Add to Cart

ARID5B Polyclonal Antibody

ARID5B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARID5B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene ARID5B
Gene Accession No. Q14865
Gene Alias ARID domain-containing protein 5B; ARID5B; AT rich interactive domain 5B (MRF1-like); AT-rich interaction domain 5B; AT-rich interactive domain 5B (MRF1-like); AT-rich interactive domain-containing protein 5B; DESRT; Modulator recognition factor 2; modulator recognition factor 2 (MRF2); MRF1-like protein; MRF2; MRF-2
Gene Symbols ARID5B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human ARID5B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.