Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARIH1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001162
Description
ARIH1 Polyclonal specifically detects ARIH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
ARIH1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ARI-1, ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1(Drosophila), ariadne, Drosophila, homolog of, ARIMOP-6, FLJ20329, FLJ93118, H7-AP2, HARI, Monocyte protein 6, MOP6, protein ariadne-1 homolog, UbcH7-binding protein, UBCH7BPariadne (Drosophila) homolog, ubiquitin-conjugating enzyme E2-binding protein1,HHARIDKFZp686O13120, UbcM4-interacting protein, Ubiquitin-conjugating enzyme E2-binding protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
25820 | |
Human | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.2 - 1 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_005735. | |
ARIH1 | |
Peptide with sequence C-HEGYEKDLWEYIED corresponding to C-Terminus according to NP_005735.2. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction