Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARIH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551885 Supplier Invitrogen™ Supplier No. PA551885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQHMVECIRE VNEVIQNPAT ITRILLSHFN WDKEKLMERY FDGNLEKLFA ECHVINPSKK SRTRQMNTRS SAQDMPCQIC YLNYPNSYFT GLECGHKFCM QCWSEYLTTK IMEEGMGQTI SCPAHGCDIL VDDNTVMRLI TDSKVK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

E3 ubiquitin-protein ligase, which catalyzes ubiquitination of target proteins together with ubiquitin-conjugating enzyme E2 UBE2L3 (PubMed:15236971, PubMed:21532592, PubMed:24076655, PubMed:27565346, PubMed:23707686). Acts as an atypical E3 ubiquitin-protein ligase by working together with cullin-RING ubiquitin ligase (CRL) complexes and initiating ubiquitination of CRL substrates: associates with CRL complexes and specifically mediates addition of the first ubiquitin on CRLs targets (PubMed:27565346). The initial ubiquitin is then elongated by CDC34/UBE2R1 and UBE2R2 (PubMed:27565346). E3 ubiquitin-protein ligase activity is activated upon binding to neddylated cullin-RING ubiquitin ligase complexes (PubMed:24076655, PubMed:27565346). Plays a role in protein translation in response to DNA damage by mediating ubiquitination of EIF4E2, the consequences of EIF4E2 ubiquitination are however unclear (PubMed:25624349). According to a report, EIF4E2 ubiquitination leads to promote EIF4E2 cap-binding and protein translation arrest (PubMed:25624349). According to another report EIF4E2 ubiquitination leads to its subsequent degradation (PubMed:14623119). Acts as the ligase involved in ISGylation of EIF4E2 (PubMed:17289916). In vitro, controls the degradation of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex member SUN2 and may therefore have a role in the formation and localization of the LINC complex, and as a consequence, nuclear subcellular localization and nuclear morphology (PubMed:29689197). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARIH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARIH1
Gene Accession No. Q9Y4X5
Gene Alias ARI; ARI-1; ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1; ariadne RBR E3 ubiquitin protein ligase 1; ariadne, Drosophila, homolog of; ARIH1; DKFZp686O13120; E3 ubiquitin-protein ligase ARIH1; FLJ20329; FLJ93118; H7-AP2; HARI; HHARI; HUSSY-27; Monocyte protein 6; MOP6; MOP-6; Protein ariadne-1 homolog; ubcH7-binding protein; UBCH7BP; ubcM4-interacting protein; Ubiquitin-conjugating enzyme E2-binding protein 1
Gene Symbols ARIH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARIH1. Recombinant protein control fragment (Product #RP-102072).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.