Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARIH2 Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
SDP

Catalog No. 502543862
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3862 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3862 Supplier OriGene Supplier No. CF811978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARIH2 Monoclonal antibody specifically detects ARIH2 in Human samples. It is validated for Western Blot

Specifications

Antigen ARIH2
Applications Western Blot
Classification Monoclonal
Clone OTI1B2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene LAMP1
Gene Accession No. P11279
Gene Alias AI843547; all-trans retinoic acid inducible RING finger; ARI2; ARI-2; ariadne 2; ariadne homolog 2; ariadne homolog 2 (Drosophila); ariadne RBR E3 ubiquitin protein ligase 2; ARIH2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; HT005; OTTHUMP00000210388; protein ariadne-2 homolog; Protein ariadne-2-like protein-like protein; RING-type E3 ubiquitin transferase ARIH2; Triad1; Triad1 protein; UbcM4-interacting protein 48; Uip48
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 29-382 of human LAMP1 (NP_005552) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3916
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.