Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARIH2 Monoclonal Antibody (OTI7D5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811977

Description
E3 ubiquitin-protein ligase mediating ′Lys-48′-and ′Lys-63′-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.Specifications
ARIH2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95376 | |
ARIH2 | |
Human recombit protein fragment corresponding to amino acids 201-493 of human ARIH2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI7D5 | |
Unconjugated | |
ARIH2 | |
ARI2, TRIAD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10425 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction