Learn More
ARIH2 Mouse anti-Human, Clone: OTI3F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804094

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
E3 ubiquitin-protein ligase mediating ′Lys-48′-and ′Lys-63′-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.Specifications
ARIH2 | |
Monoclonal | |
Unconjugated | |
ARIH2 | |
ARI2, TRIAD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10425 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95376 | |
ARIH2 | |
Human recombit protein fragment corresponding to amino acids 201-493 of human ARIH2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.