Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARIH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528446
Description
Recommended positive controls: HeLa, mouse muscle. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 ubiquitin-protein ligase mediating 'Lys-48'-and 'Lys-63'-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.
Specifications
| ARIH2 | |
| Polyclonal | |
| Unconjugated | |
| Arih2 | |
| AI843547; all-trans retinoic acid inducible RING finger; ARI2; ARI-2; ariadne 2; ariadne homolog 2; ariadne homolog 2 (Drosophila); ariadne RBR E3 ubiquitin protein ligase 2; ARIH2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; HT005; OTTHUMP00000210388; protein ariadne-2 homolog; Protein ariadne-2-like protein-like protein; RING-type E3 ubiquitin transferase ARIH2; Triad1; Triad1 protein; UbcM4-interacting protein 48; Uip48 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10425, 23807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O95376, Q9Z1K6 | |
| Arih2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ARIH2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction