Learn More
Invitrogen™ ARIH2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ARIH2.
Specifications
Specifications
| Antigen | ARIH2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Arih2 |
| Gene Accession No. | O95376, Q9Z1K6 |
| Gene Alias | AI843547; all-trans retinoic acid inducible RING finger; ARI2; ARI-2; ariadne 2; ariadne homolog 2; ariadne homolog 2 (Drosophila); ariadne RBR E3 ubiquitin protein ligase 2; ARIH2; E3 ubiquitin-protein ligase ARIH2; FLJ10938; FLJ33921; HT005; OTTHUMP00000210388; protein ariadne-2 homolog; Protein ariadne-2-like protein-like protein; RING-type E3 ubiquitin transferase ARIH2; Triad1; Triad1 protein; UbcM4-interacting protein 48; Uip48 |
| Gene Symbols | Arih2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.