Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARIH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729904 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9904 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9904 Supplier Proteintech Group Inc Supplier No. 150061AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E3 ubiquitin-protein ligase mediating ′Lys-48′-and ′Lys-63′-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.

Specifications

Antigen ARIH2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARIH2
Gene Accession No. O95376, Q9Z1K6
Gene Alias ARI 2, ARI2, ariadne homolog 2 (Drosophila), ARIH2, Protein ariadne 2 homolog, TRIAD1, Triad1 protein
Gene Symbols Arih2
Host Species Rabbit
Immunogen ARIH2 Fusion Protein Ag6507
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10425, 23807, 316005
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.