Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ comfort I ASTM 1, 3 Combined Water purification Systems with UV Lamp
GSA_VAGREENER_CHOICE

Catalog No. 15555515 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
16 L/hr.
8 L/hr.
Mounting:
Benchtop
Wall
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Mounting
15-555-515 8 L/hr. Wall
15-555-511 8 L/hr. Benchtop
15-555-513 16 L/hr. Benchtop
15-555-517 16 L/hr. Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-555-515 Supplier Sartorius Supplier No. H2OI1UVB
Add to Cart
Add to Cart
Request a Quote

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.

  • Unique cartridge kit for production of highest quality ultrapure water
  • Innovative iJust technology optimizes product water quality
  • Touch display with intuitive menu navigation
  • Compact design with SD card slot
  • Reproducible and reliable results
  • Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
  • Available as bench-top system for every laboratory workstation
  • Wall-mounted with integrated wall bracket

Benefits

  • Optimized, economic water consumption
  • Premium product-water quality at all times
  • Guaranteed longer life of downstream ultrapure water systems

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 8 L/hr.
Product Water Type Type 1 and 3
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Feedwater Pressure 2.0 to 6.9 bar
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, Photometry, Buffer solutions, Feed water for laboratory devices, such as autoclaves, glassware washers etc
Humidity Range 80%
Mounting Wall
Particle Size <1mL
Purification Method Adsorption by spherical activated carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration
Turbidity <1 NTU
Width (English) 17 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.7 in.
Depth (Metric) 47.6 cm
Weight (English) 68.3 lb. (Operating); 50.7 lb. (Empty)
Weight (Metric) Approx. 31 kg (Operating); 23 kg (Empty)
Electrical Requirements 100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.)
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Water Purification System
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.