Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ comfort II ASTM 1, 2 Combined Water Purification Systems w/o UV Lamp, TOC Monitor
GSA_VAGREENER_CHOICE

Catalog No. p-5013620
Click to view available options
Flow Rate:
10 L/hr.
5 L/hr.
Mounting:
Benchtop
Wall

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

  • Equipped with latest EDI technology and unique cartridge kit
  • Innovative iJust technology optimizes product water quality
  • Touch display with intuitive menu navigation
  • Compact design with SD card slot
  • Performs electrochemical deionization in third purification stage
  • Reliable and safe removal of contamination contained in feed water
  • Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2

Benefits

  • Optimized, economic water consumption
  • Premium product-water quality at all times
  • Guaranteed longer life of downstream ultrapure water systems

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 5 L/hr.
Product Water Type Type 1 and 2
Remote Dispenser No
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring < 5 ppb
Total Organic Carbon <5 ppb
Type Water Purification System
Feedwater Pressure 2.0 to 6.9 bar
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, LISA, RIA
Humidity Range 80%
Mounting Benchtop
Particle Size <1mL
Purification Method Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration
Turbidity <1 NTU
Width (English) 17 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.7 in.
Depth (Metric) 47.6 cm
Weight (English) 61.7 lb. (Empty); 79.3 lb. (Operating)
Weight (Metric) 28 kg (Empty); 36 kg (Operating)
Electrical Requirements 100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.