Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius arium™ comfort II Combined System (TYPE 1 - Ultrapure Water and TYPE 2 - EDI Water)
GSA_VAGREENER_CHOICE

Catalog No. 15555518 Shop All Sartorius Products
Change view
Click to view available options
Bacterial Content:
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bacterial Content
15-555-518 <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-555-518 Supplier Sartorius Supplier No. H2OII1TOCT
Add to Cart
Add to Cart
Request a Quote

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

  • Flexible, space-saving twin technology
  • Efficient iJust technology
  • Uses disposable bagtank technology with dispense gun connections
  • Built-in TOC monitor analyzes and displays actual TOC values
  • Integrated UV lamp, 185/254nm, reduce the TOC less-than-or-equal-to 2ppb
  • Convenient SD card slot
  • User-friendly menu navigation
  • Meet customerapostrophes lab requirements and saves maintenance time
  • Safe operation
  • Reliable TOC reduction
  • Supports reliable recording of important data

To install arium comfort II as a built-in unit, please order the corresponding bench-top system and a conversion kit H2O-ACK-D

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Conductivity 0.055 Product water, Less than1500 feed water
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 2 L/min.
Product Water Type Type 1 and 2
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Feedwater Temperature (English) 35.6°F to 86°F
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) HPLC, AAS, Ion chromatography, Photometry, Microbiological media and reagents, Histology, ELISA, RIA, Buffers and pH solutions Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths
Humidity Range 80%
Includes UV lamp, TOC monitor
Particle Size <1/mL
pH Range 4 to 10
Storage Requirements 5°C to 45°C
Temperature Range (English) 35.6°F to 95°F
Temperature Range (Metric) 2°C to 35°C
Tolerance <4
Turbidity <1
Width (English) 17.12 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.74 in.
Depth (Metric) 47.6 cm
Weight (English) 79.36 lb.
Weight (Metric) 36 kg
Electrical Requirements 100/240 V 50/60 Hz
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Water Purification System
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.