Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Arium™ Mini Essential Set for Production of ASTM Type 1 Ultrapure Water with UV
GSA_VAGREENER_CHOICE

Catalog No. 14558732 Shop All Sartorius Products
Change view
Click to view available options
Bacterial Content:
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bacterial Content
14-558-732 <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-732 Supplier Sartorius Supplier No. H2OMUUVSETUS
Add to Cart
Add to Cart
Request a Quote

Compact Arium™ mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 liters per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.

  • Directly connects to a pretreated water supply line (RO, DI, EDI)
  • Reliable: Delivers consistently high water quality for reliable and reproducible results
  • Intuitive: Touch-activated color display with direct access to all important dispensing options
  • Innovative: saves time-intensive tank cleaning
  • Compact: Space-saving, with a width of only 28 cm. UV Lamp for Critical Analyses (185, 254 nm)
  • Optimal temperature gradient due to horizontal installation
  • Degradation of total organic carbons (TOCs) by irradiation (< 5 ppb)
  • Inactivates microorganisms by damaging their DNA.
  • Set includes: Arium™ Scientific Kit, H2O-S-Pack Fisher Scientific Catalog Number: 14-559-045 ,Arium™ Sterile Filter, 5441307H4--CE Fisher Scientific Catalog Number: 14-557-601

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Conductivity < 100 μS/cm, compensated to 25°C
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring Water pretreated by reverse osmosis, distillation or deionization
Feedwater Source Pretreated
Flow Rate Up to 1.0 L/min.
Product Water Type Type 1
Remote Dispenser No
Resistivity 18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring < 5 ppb
Total Organic Carbon <10 ppb
Display Touch activated
Feedwater Temperature (Metric) 0 to 6.9 bar; (0 to approx. 100 psi); recommended > 2 bar (>29 psi)
For Use With (Equipment) arium mini accessories
Dimensions (D x W x H) Interior 20.8 x 11 x 20 in.
Height (Metric) 509 mm
Electrical Requirements 100/240 VAC 50/60 Hz 2.0 A max
Voltage 100/240 V
Amperage Input supply voltage VDC +24 (±10%) Current consumption, max. A 3.0
Daily Output 10 L
Endotoxin Concentration <0.001 EU/mL
Product Type Lab Water System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.