Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Arium™ Mini Extend Ultrapure Water System

Catalog No. 01673426 Shop All Sartorius Products
Change view
Click to view available options
Model:
Arium Mini Essential with UV
Arium Mini Essential without UV
Arium Mini Plus with UV
Arium Mini Plus without UV
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-673-426 Arium Mini Essential with UV
01-673-429 Arium Mini Plus without UV
01-673-428 Arium Mini Plus with UV
01-673-427 Arium Mini Essential without UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-673-426 Supplier Sartorius Supplier No. H2OMURUVT
Add to Cart
Add to Cart
Request a Quote

Compact Arium™ Mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 L/day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.

  • Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
  • Compact: Space-saving with a width of only 28 cm (+ 9 cm for versions with attached dispensing unit)
  • Intuitive: Color touch display, with direct access to all important information and dispensing options
  • Innovative: Depending on the type of system, our unique Bagtank technology saves time as no intensive tank cleaning with hazardous substances is required
  • Reliable: Delivers consistently high water quality for reliable and reproducible results
TRUSTED_SUSTAINABILITY

Specifications

Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Distilled, DI, RO, EDI
Flow Rate 1 L/min.
Product Water Type Type 1
Remote Dispenser Optional
Resistivity 18.2 MΩ x cm
TOC Monitor with UV Intensity Monitoring < 5 ppb
Total Organic Carbon ≤5 ppb
Model Arium Mini Essential with UV
Daily Output 1 to 10 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Water Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.