Learn More
Description
- Height adjustable display/dispense station for dispensing Type 1 ultrapure water
- Compact design for easy and space-saving integration into lab environment
- Intuitive touch-activated color display with direct access to important functions
- Flexible and stepless height adjustment to fill different sized containers and beakers
- Ergonomic design can be optimized for right or left hand operation
- Accurate volume dispense for fast and precise buffer and sample prep
- Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
- Display/dispense unit with stand
- Power supply including:
- IEC connector
- Connection set
For intended use with:
- Arium™ Bagtank
- Arium™ Pro
- Arium™ Comfort
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Feed Water Monitoring | No |
| Feedwater Source | Ultrapure |
| Flow Rate | Up to 2 L/min. |
| Product Water Type | Ultrapure (Type 1) |
| TOC Monitor with UV Intensity Monitoring | No |
| Type | Remote Dispenser |
| Display | Touch display |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
