Learn More
Sartorius™ Arium™ SterilePlus PROMO
Catalog No. 14557601PM
Click to view available options
For Use With (Equipment):
Arium™ Pro, Mini, Comfort, Remote Dispenser, Smart Station
Sterile final filter for sterile and particle-free water dispensing.
  • Consists of combined 0.45 μm and 0.2 μm polyethersulfone membrane
  • For intended use with: Arium™ Mini, Mini Essential, Mini Plus, Pro, Pro DI, Pro UV, Pro UF, Pro VF, Comfort I, Comfort II, Remote Dispenser, Smart Station
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium™ Pro, Mini, Comfort, Remote Dispenser, Smart Station
Material PES
Pore Size (Metric) 0.45 μm and 0.2 μm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.