Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528776 Supplier Invitrogen™ Supplier No. PA528776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HUVEC. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the ARL (ADP-ribosylation factor-like) family of proteins, which are structurally related to ADP-ribosylation factors (ARFs). ARFs, described as activators of cholera toxin (CT) ADP-ribosyltransferase activity, regulate intracellular vesicular membrane trafficking, and stimulate a phospholipase D (PLD) isoform. Although, ARL proteins were initially thought not to activate CT or PLD, later work showed that they are weak stimulators of PLD and CT in a phospholipid dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ARL1
Gene Accession No. P40616
Gene Alias 2310008D22Rik; ADP ribosylation factor like GTPase 1; ADP-ribosylation factor like GTPase 1; ADP-ribosylation factor-like 1; ADP-ribosylation factor-like protein 1; ADP-ribosylation-like 1; Arf7; ARFL1; Arl1; AU019749
Gene Symbols ARL1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ARL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.