Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561412 Supplier Invitrogen™ Supplier No. PA561412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGKSTFLRVL SGKPPLEGHI PTWGFNSVRL PTKDFEVDLL EIGGSQNLRF YWKEFVSEVD VLVFVVDSAD RLRLPWARQE LHKLLDKDPD LPVV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

ARL10 (ADP Ribosylation Factor Like GTPase 10) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include GTP binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARL10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL10
Gene Accession No. Q8N8L6
Gene Alias ADP ribosylation factor like GTPase 10; ADP-ribosylation factor like GTPase 10; ADP-ribosylation factor-like 10A; ADP-ribosylation factor-like membrane-associated protein; ADP-ribosylation factor-like protein 10; ARL10; ARL10A
Gene Symbols ARL10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARL10. Recombinant protein control fragment (Product #RP-99919).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285598
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.