Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL11 Antibody (OTI1A5), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272348JF646
Description
ARL11 Monoclonal specifically detects ARL11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ARL11 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARL11 | |
Full length recombinant protein of human ARL11 (NP_612459) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ADP-ribosylation factor-like 11, ADP-ribosylation factor-like protein 11, ADP-ribosylation factor-like tumor suppressor protein 1, ARLTS1FLJ33930, MGC17429 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
115761.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction