Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARL11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100781
Description
Antibody detects endogenous levels of total ARL11.
This gene encodes a tumor suppressor related to the ADP-ribosylation factor (ARF) family of proteins. The encoded protein may play a role in apoptosis in a caspase-dependent manner. Polymorphisms in this gene have been associated with some familial cancers.
Specifications
ARL11 | |
Polyclonal | |
Unconjugated | |
ARL11 | |
ADP ribosylation factor like GTPase 11; ADP-ribosylation factor like GTPase 11; ADP-ribosylation factor-like 11; ADP-ribosylation factor-like protein 11; ADP-ribosylation factor-like tumor suppressor protein 1; ARL11; ARLTS1; C730007L20Rik | |
Rabbit | |
Affinity chromatography | |
RUO | |
115761, 364396 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5BK71, Q969Q4 | |
ARL11 | |
A synthesized peptide derived from human ARL11(Accession Q969Q4), corresponding to amino acid residues L70-L120. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction